Gene Transcripts

Gene Symbol
SUPT6H
Entrez Gene ID
6830
Gene Name
SPT6 homolog, histone chaperone
Gene Aliases
SPT6, SPT6H, emb-5
Chromosome Location
Chr. 17: 28662183 - 28702684 on Build GRCh38
UniGene ID
Hs.250429
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001320755.1 NP_001307684.1 33, 34 4734
NM_003170.4 NP_003161.2 32, 33 4632
XM_017024978.1 XP_016880467.1 34, 35 4793
XM_017024979.1 XP_016880468.1 32, 33 4870
XM_017024980.1 XP_016880469.1 31, 32 4564
GenBank mRNA AF070532.1 - 1, 2 81
AL833607.1 - 33, 34 4735
BC003692.1 - 4, 5 624
BC003696.1 - 17, 18 2507
BC017105.1 - 12, 13 1775
BC073963.1 - 5, 6 931
BC136522.1 - 32, 33 4544 9020512
BC136524.1 - 32, 33 4544 9020514
BC150268.1 - 32, 33 4546 9007230
D79984.1 - 32, 33 4546

View Full Product Details