Gene Transcripts
- Gene Symbol
- Arid4b
- Entrez Gene ID
- 84481
- Gene Name
- AT rich interactive domain 4B (Rbp1 like)
- Gene Aliases
- Bcaa
- Chromosome Location
- Chr. 17: 53831353 - 53955920 on Build Rnor_6.0
- UniGene ID
- Rn.47249
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_053421.1 |
NP_445873.1
|
18 |
2264 |
|
GenBank mRNA |
AF245512.1 |
- |
18 |
2264 |
|
View Full Product Details