Gene Transcripts

Gene Symbol
Sbk1
Entrez Gene ID
113907
Gene Name
SH3 domain binding kinase 1
Gene Aliases
Sbk
Chromosome Location
Chr. 1: 197659113 - 197681031 on Build Rnor_6.0
UniGene ID
Rn.35152
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_147135.2 NP_671476.1 3 745
XM_008759853.1 XP_008758075.1 3 752
XM_008759854.1 XP_008758076.1 4 1080
GenBank mRNA AB010154.1 - 3 457
BC131843.1 - 3 745 8363395

Gene Symbol
LOC103690016
Entrez Gene ID
103690016
Gene Name
serine/threonine-protein kinase SBK1
Gene Aliases
Chromosome Location
Chr. 1: 67368372 - 67390254 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008758862.1 XP_008757084.1 3 793
XM_008758863.1 XP_008757085.1 3 750
XM_008758864.1 XP_008757086.1 4 1054

View Full Product Details