Gene Transcripts

Gene Symbol
Gpx5
Entrez Gene ID
113919
Gene Name
glutathione peroxidase 5
Gene Aliases
Chromosome Location
Chr. 17: 45508657 - 45518686 on Build Rnor_6.0
UniGene ID
Rn.218434
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001105738.1 NP_001099208.1 359
GenBank mRNA X62404.1 - 404

View Full Product Details