Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001270488.1 | NP_001257417.1 | 4 | 589 | |
NM_005643.3 | NP_005634.1 | 5 | 686 | ||
XM_011514827.2 | XP_011513129.1 | 5 | 1076 | ||
GenBank mRNA | AF118094.1 | - | 5 | 426 | |
AK302226.1 | - | 4 | 560 | ||
AK313475.1 | - | 5 | 627 | ||
BC021972.2 | - | 5 | 642 | 3935882 | |
BT019404.1 | - | 5 | 572 | ||
BT019405.1 | - | 5 | 572 | ||
BX538195.1 | - | 2 | 1954 | ||
BX647568.1 | - | 3 | 2857 | ||
CR542068.1 | - | 5 | 572 | ||
D63705.1 | - | 5 | 655 | ||
X83928.1 | - | 5 | 664 |