Gene Transcripts

Gene Symbol
Hcn3
Entrez Gene ID
114245
Gene Name
hyperpolarization-activated cyclic nucleotide-gated potassium channel 3
Gene Aliases
Chromosome Location
Chr. 2: 188458851 - 188471916 on Build Rnor_6.0
UniGene ID
Rn.48801
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053685.1 NP_446137.1 2 636
GenBank mRNA AF155165.1 - 1 363
AF247452.1 - 2 636

View Full Product Details