Gene Transcripts

Gene Symbol
Wnt2b
Entrez Gene ID
116466
Gene Name
wingless-type MMTV integration site family, member 2B
Gene Aliases
Chromosome Location
Chr. 2: 207440886 - 207455258 on Build Rnor_6.0
UniGene ID
Rn.17219
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001191848.1 NP_001178777.1 3 584
GenBank mRNA AF204873.1 - 2 344

View Full Product Details