Gene Transcripts

Gene Symbol
Pcdha13
Entrez Gene ID
116742
Gene Name
protocadherin alpha 13
Gene Aliases
Cnr5
Chromosome Location
Chr. 18: 30036887 - 30215897 on Build Rnor_6.0
UniGene ID
Rn.22308
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053934.2 NP_446386.2 1 751
GenBank mRNA AB045586.1 - 1 29
AB113396.1 - 1 751
AY573975.1 - 1 751

Gene Symbol
LOC317356
Entrez Gene ID
317356
Gene Name
similar to protocadherin alpha 13
Gene Aliases
Chromosome Location
Chr. X: 13709703 - 13713872 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006256688.1 XP_006256750.1 2 175

View Full Product Details