Gene Transcripts

Gene Symbol
Cdh3
Entrez Gene ID
116777
Gene Name
cadherin 3
Gene Aliases
Chromosome Location
Chr. 19: 38668953 - 38719823 on Build Rnor_6.0
UniGene ID
Rn.105829
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006255559.2 XP_006255621.1 14, 15 2681
GenBank mRNA AF177683.1 - 12

View Full Product Details