Gene Transcripts

Gene Symbol
Rhbdl1
Entrez Gene ID
117025
Gene Name
rhomboid, veinlet-like 1 (Drosophila)
Gene Aliases
Rhbdl
Chromosome Location
Chr. 10: 15201503 - 15205885 on Build Rnor_6.0
UniGene ID
Rn.94890
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001191822.1 NP_001178751.1 2 141
XM_008767537.1 XP_008765759.1 2 1883

View Full Product Details