Gene Transcripts

Gene Symbol
HNF1A
Entrez Gene ID
6927
Gene Name
HNF1 homeobox A
Gene Aliases
HNF-1A, HNF1, IDDM20, LFB1, MODY3, TCF-1, TCF1
Chromosome Location
Chr. 12: 120978515 - 121002512 on Build GRCh38
UniGene ID
Hs.654455
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000545.6 NP_000536.5 6 1476
NM_001306179.1 NP_001293108.1 6 1476
XM_005253931.3 XP_005253988.1 6 1529
GenBank mRNA AK298422.1 - 6 1350
BC104908.1 - 6 1295 8143911
BC104910.1 - 6 1295 8143913
BC143483.1 - 6 1296
HM116541.1 - 6 1418
HM116542.1 - 6 1092
HM116543.1 - 4 881
HM116545.1 - 3 405
HM116547.1 - 6 1418
HM116552.1 - 7 1035
HM116557.1 - 5 1206
HM116558.1 - 5 1151
HM116559.1 - 2 346
HM116560.1 - 3 270
HM116563.1 - 2 282
HM449088.1 - 6 1298
HM449089.1 - 5 1098
M57732.1 - 6 1298
X71346.1 - 6 1471
X71347.1 - 6 1406

View Full Product Details