Gene Transcripts
- Gene Symbol
- Smu1
- Entrez Gene ID
- 117541
- Gene Name
- smu-1 suppressor of mec-8 and unc-52 homolog (C. elegans)
- Gene Aliases
- Bwd, Smu-1
- Chromosome Location
- Chr. 5: 57042743 - 57061313 on Build Rnor_6.0
- UniGene ID
- Rn.54978
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_057195.1 |
NP_476543.1
|
6, 7 |
779 |
|
GenBank mRNA |
AY029526.1 |
- |
6, 7 |
779 |
|
View Full Product Details