Gene Transcripts

Gene Symbol
Cdk7
Entrez Gene ID
171150
Gene Name
cyclin-dependent kinase 7
Gene Aliases
Chromosome Location
Chr. 2: 30710642 - 30735520 on Build Rnor_6.0
UniGene ID
Rn.98896
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008760700.1 XP_008758922.1 11 1019
GenBank mRNA X83579.1 - 11 865

View Full Product Details