Gene Transcripts

Gene Symbol
Cox8a
Entrez Gene ID
171335
Gene Name
cytochrome c oxidase subunit VIIIa
Gene Aliases
Chromosome Location
Chr. 1: 222466575 - 222468896 on Build Rnor_6.0
UniGene ID
Rn.2270
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_134345.1 NP_599172.1 2 362
GenBank mRNA BC126066.1 - 2 329
L48209.1 - 2 280
X06146.1 - 207

View Full Product Details