Gene Transcripts
- Gene Symbol
- Slc22a25
- Entrez Gene ID
- 192273
- Gene Name
- solute carrier family 22, member 25
- Gene Aliases
- Slc22a9, Ust1, Ust1r
- Chromosome Location
- Chr. 1: 224596886 - 224698512 on Build Rnor_6.0
- UniGene ID
- Rn.31066
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_138908.2 |
NP_620263.1
|
7 |
1174 |
|
GenBank mRNA |
Y09945.1 |
- |
7 |
1176 |
|
View Full Product Details