Gene Transcripts

Gene Symbol
LOC246267
Entrez Gene ID
246267
Gene Name
resection-induced TPI (rs11)
Gene Aliases
Chromosome Location
Chr. : - on Build
UniGene ID
Rn.127819
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_144751.1 NP_653352.1 160
GenBank mRNA AF007890.1 - 160

View Full Product Details