Gene Transcripts

Gene Symbol
Cyp4f5
Entrez Gene ID
286905
Gene Name
cytochrome P450, family 4, subfamily f, polypeptide 5
Gene Aliases
Chromosome Location
Chr. 7: 14559806 - 14574345 on Build Rnor_6.0
UniGene ID
Rn.10171
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173124.1 NP_775147.1 1298
XM_006241051.2 XP_006241113.1 10 1367
XM_008765157.1 XP_008763379.1 9 1315
XM_008765158.1 XP_008763380.1 10 1369
GenBank mRNA BC129085.1 - 10 1259
U39207.1 - 1298

Gene Symbol
Cyp4f37
Entrez Gene ID
691312
Gene Name
cytochrome P450, family 4, subfamily f, polypeptide 37
Gene Aliases
Chromosome Location
Chr. 7: 14589520 - 14614869 on Build Rnor_6.0
UniGene ID
Rn.10171
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001271352.1 NP_001258281.1 12 1756

Gene Symbol
LOC100912391
Entrez Gene ID
100912391
Gene Name
cytochrome P450 4F5-like
Gene Aliases
Chromosome Location
Chr. 7: 14672524 - 14679558 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006241095.2 XP_006241157.1 5 656

Gene Symbol
LOC100912471
Entrez Gene ID
100912471
Gene Name
cytochrome P450 4F5-like
Gene Aliases
Chromosome Location
Chr. 7: 14760053 - 14830055 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006241096.2 XP_006241158.1 2192
XM_006241097.2 XP_006241159.1 8 934

Gene Symbol
LOC691320
Entrez Gene ID
691320
Gene Name
similar to cytochrome P450 4F1
Gene Aliases
Chromosome Location
Chr. 7: 13881303 - 13889392 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006241044.2 XP_006241106.1 5 2192
XM_006241045.1 XP_006241107.1 7 901

View Full Product Details