Gene Transcripts

Gene Symbol
Tox4
Entrez Gene ID
286990
Gene Name
TOX high mobility group box family member 4
Gene Aliases
RGD708449
Chromosome Location
Chr. 15: 28696378 - 28709960 on Build Rnor_6.0
UniGene ID
Rn.22195
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173324.2 NP_775446.2 4 420
GenBank mRNA AF267197.1 - 3 372
BC081696.1 - 4 420 7112549
FQ210274.1 - 4 428
FQ212245.1 - 4 428

View Full Product Details