Gene Transcripts

Gene Symbol
Zmynd15
Entrez Gene ID
287457
Gene Name
zinc finger, MYND-type containing 15
Gene Aliases
Chromosome Location
Chr. 10: 57064482 - 57071096 on Build Rnor_6.0
UniGene ID
Rn.163126
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001105800.1 NP_001099270.1 11, 12 2352
XM_006246661.2 XP_006246723.1 10, 11 1867
XM_006246662.2 XP_006246724.1 10, 11 1854

View Full Product Details