Gene Transcripts

Gene Symbol
Ift20
Entrez Gene ID
287541
Gene Name
intraflagellar transport 20
Gene Aliases
RGD1309400
Chromosome Location
Chr. 10: 65805814 - 65811353 on Build Rnor_6.0
UniGene ID
Rn.8370
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001105815.1 NP_001099285.1 5 707
XM_006246935.2 XP_006246997.1 4 521

View Full Product Details