Gene Transcripts

Gene Symbol
Pms2
Entrez Gene ID
288479
Gene Name
postmeiotic segregation increased 2 (S. cerevisiae)
Gene Aliases
Chromosome Location
Chr. 12: 12714394 - 12738641 on Build Rnor_6.0
UniGene ID
Rn.102072
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001105908.1 NP_001099378.1 5 498
XM_006248862.2 XP_006248924.1 5 493
GenBank mRNA BC160858.1 - 5 432

View Full Product Details