Gene Transcripts

Gene Symbol
Ksr2
Entrez Gene ID
288691
Gene Name
kinase suppressor of ras 2
Gene Aliases
RGD1562674
Chromosome Location
Chr. 12: 44531376 - 44911824 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006249485.2 XP_006249547.1 9 2141

View Full Product Details