Gene Transcripts

Gene Symbol
Foxn4
Entrez Gene ID
288736
Gene Name
forkhead box N4
Gene Aliases
Chromosome Location
Chr. 12: 48101012 - 48121281 on Build Rnor_6.0
UniGene ID
Rn.154105
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001105935.1 NP_001099405.1 10 1185
XM_006249493.2 XP_006249555.1 10 1726

View Full Product Details