Gene Transcripts

Gene Symbol
Mrps14
Entrez Gene ID
289143
Gene Name
mitochondrial ribosomal protein S14
Gene Aliases
Chromosome Location
Chr. 13: 77940454 - 77946201 on Build Rnor_6.0
UniGene ID
Rn.67
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001105963.2 NP_001099433.1 2 189
XM_006250098.2 XP_006250160.1 2 306
GenBank mRNA BC166895.1 - 2 189
FQ214536.1 - 2 173

View Full Product Details