Gene Transcripts

Gene Symbol
TRPC3
Entrez Gene ID
7222
Gene Name
transient receptor potential cation channel subfamily C member 3
Gene Aliases
SCA41, TRP3
Chromosome Location
Chr. 4: 121879028 - 121952060 on Build GRCh38
UniGene ID
Hs.150981
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001130698.1 NP_001124170.1 7 2016
NM_003305.2 NP_003296.1 6 1798
XM_011532217.2 XP_011530519.1 7 2322
XM_011532218.2 XP_011530520.1 7 2322
XM_017008578.1 XP_016864067.1 7 2322
XM_017008579.1 XP_016864068.1 7 2322
GenBank mRNA AB255423.1 - 5 1558
AB255424.1 - 6 1723
AY865574.1 - 7 2016
BC093682.1 - 6 1798 7939527
BC093684.1 - 6 1798 7939529
Y13758.1 - 6 2147

View Full Product Details