Gene Transcripts

Gene Symbol
Lefty2
Entrez Gene ID
289316
Gene Name
Left-right determination factor 2
Gene Aliases
Ebaf2
Chromosome Location
Chr. 13: 99173484 - 99178037 on Build Rnor_6.0
UniGene ID
Rn.215726
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001007556.1 NP_001007557.1 3, 4 798
GenBank mRNA AY758558.1 - 3, 4 798

View Full Product Details