Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_003311.3 | NP_003302.1 | 2 | 649 | |
GenBank mRNA | AF001294.1 | - | 2 | 649 | |
AF019953.1 | - | 2 | 610 | ||
AF035444.1 | - | 2 | 649 | ||
AK223027.1 | - | 2 | 649 | ||
BC005034.1 | - | 2 | 639 | 3954346 |