Gene Transcripts

Gene Symbol
Mustn1
Entrez Gene ID
290553
Gene Name
musculoskeletal, embryonic nuclear protein 1
Gene Aliases
Mustang
Chromosome Location
Chr. 16: 6962737 - 6964987 on Build Rnor_6.0
UniGene ID
Rn.203378
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_181368.3 NP_852033.1 2 394
GenBank mRNA AY254906.2 - 2 394
BC158570.1 - 2 409 9092791

View Full Product Details