Gene Transcripts

Gene Symbol
Klhl26
Entrez Gene ID
290657
Gene Name
kelch-like family member 26
Gene Aliases
RGD1312034
Chromosome Location
Chr. 16: 20704807 - 20729882 on Build Rnor_6.0
UniGene ID
Rn.23695
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106075.1 NP_001099545.1 3 429

View Full Product Details