Gene Transcripts

Gene Symbol
XIAP
Entrez Gene ID
331
Gene Name
X-linked inhibitor of apoptosis
Gene Aliases
API3, BIRC4, IAP-3, ILP1, MIHA, XLP2, hIAP-3, hIAP3
Chromosome Location
Chr. X: 123859812 - 123913979 on Build GRCh38
UniGene ID
Hs.356076
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001167.3 NP_001158.2 2 991
NM_001204401.1 NP_001191330.1 2 958
XM_006724754.2 XP_006724817.1 3 1071
XM_011531329.2 XP_011529631.1 2 961
GenBank mRNA AK313883.1 - 2 958
BC032729.1 - 2 910 5532247
U32974.1 - 2 913

View Full Product Details