Gene Transcripts

Gene Symbol
Diras2
Entrez Gene ID
291006
Gene Name
DIRAS family, GTP-binding RAS-like 2
Gene Aliases
Chromosome Location
Chr. 17: 12737439 - 12766694 on Build Rnor_6.0
UniGene ID
Rn.214733
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001169578.1 NP_001163049.1 1 517
XM_006253671.2 XP_006253733.1 2 582
XM_008771489.1 XP_008769711.1 2 601

View Full Product Details