Gene Transcripts

Gene Symbol
Nrsn1
Entrez Gene ID
291129
Gene Name
neurensin 1
Gene Aliases
Vmp
Chromosome Location
Chr. 17: 41798767 - 41816346 on Build Rnor_6.0
UniGene ID
Rn.225156
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106109.2 NP_001099579.2 3 327
GenBank mRNA BC166881.1 - 3 327
BC166919.1 - 3 327 6919003

View Full Product Details