Gene Transcripts

Gene Symbol
Tubal3
Entrez Gene ID
291287
Gene Name
tubulin, alpha-like 3
Gene Aliases
Chromosome Location
Chr. 17: 69974272 - 69982957 on Build Rnor_6.0
UniGene ID
Rn.218522
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106118.1 NP_001099588.1 3 639

View Full Product Details