Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001282161.1 | NP_001269090.1 | 6 | 585 | |
NM_003336.3 | NP_003327.2 | 6 | 592 | ||
NM_181762.2 | NP_861427.1 | 5 | 502 | ||
GenBank mRNA | AK223045.1 | - | 6 | 494 | |
AK223064.1 | - | 6 | 491 | ||
AK297696.1 | - | 5 | 404 | ||
AK301675.1 | - | 5 | 503 | ||
AK313092.1 | - | 6 | 496 | ||
BC010175.2 | - | 6 | 485 | 3051041 | |
BC042021.1 | - | 3 | 1369 | ||
BI093289.1 | - | 5 | 292 | ||
DA068506.1 | - | 6 | 489 | ||
M74524.1 | - | 6 | 467 |