Gene Transcripts

Gene Symbol
Ftmt
Entrez Gene ID
291470
Gene Name
ferritin mitochondrial
Gene Aliases
Chromosome Location
Chr. 18: 47294292 - 47295541 on Build Rnor_6.0
UniGene ID
Rn.218703
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106136.1 NP_001099606.1 1 678

View Full Product Details