Gene Transcripts

Gene Symbol
Prmt10
Entrez Gene ID
291947
Gene Name
protein arginine methyltransferase 10 (putative)
Gene Aliases
RGD1306157
Chromosome Location
Chr. 19: 34067008 - 34105622 on Build Rnor_6.0
UniGene ID
Rn.199059
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001191599.1 NP_001178528.1 12 2463
XM_006255424.2 XP_006255486.1 12 2499

View Full Product Details