Gene Transcripts

Gene Symbol
UFD1L
Entrez Gene ID
7353
Gene Name
ubiquitin fusion degradation 1 like (yeast)
Gene Aliases
UFD1
Chromosome Location
Chr. 22: 19449941 - 19479215 on Build GRCh38
UniGene ID
Hs.474213
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001035247.2 NP_001030324.2 3, 4 309
NM_005659.6 NP_005650.2 3, 4 309
XM_011530361.2 XP_011528663.1 3, 4 435
GenBank mRNA AF141201.1 - 2, 3 179
AJ239058.1 - 2, 3 179
AK225877.1 - 3, 4 266
AK304734.1 - 3, 4 279
AK308786.1 - 3, 4 253
AK310950.1 - 3, 4 261
AY101594.1 - 3, 4 236
BC001049.2 - 3, 4 256 3507963
BC005087.1 - 3, 4 257 3830774
BM761958.1 - 2, 3 248
BX648483.1 - 3, 4 241
CB128364.1 - 3, 4 309
CR456607.1 - 3, 4 193
DB496066.1 - 3, 4 293
U64444.1 - 3, 4 257

View Full Product Details