Gene Transcripts

Gene Symbol
Fpr3
Entrez Gene ID
292413
Gene Name
formyl peptide receptor 3
Gene Aliases
RGD1566044
Chromosome Location
Chr. 1: 116152523 - 116161636 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_003748874.3 XP_003748922.1 3 439

View Full Product Details