Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001076.3 | NP_001067.2 | 1 | 457 | |
GenBank mRNA | AF180322.1 | - | 1 | 337 | |
AF548389.1 | - | 1 | 316 | ||
AK289419.1 | - | 1 | 330 | ||
BC171879.1 | - | 457 | 9054533 | ||
U06641.1 | - | 1 | 278 | ||
U08854.1 | - | 1 | 320 |