Gene Transcripts

Gene Symbol
Irgq
Entrez Gene ID
292708
Gene Name
immunity-related GTPase family, Q
Gene Aliases
RGD1309409
Chromosome Location
Chr. 1: 81395924 - 81403872 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001135742.2 NP_001129214.2 3 2124
XM_006228410.2 XP_006228472.1 2 2082

View Full Product Details