Gene Transcripts

Gene Symbol
Blvrb
Entrez Gene ID
292737
Gene Name
biliverdin reductase B
Gene Aliases
Chromosome Location
Chr. 1: 84256063 - 84273726 on Build Rnor_6.0
UniGene ID
Rn.161812
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106236.1 NP_001099706.1 4 543
XM_008759117.1 XP_008757339.1 4 464
GenBank mRNA BC168943.1 - 4 447
FQ228106.1 - 4 427
FQ230737.1 - 4 490
FQ232598.1 - 4 490

View Full Product Details