Gene Transcripts

Gene Symbol
Ankrd42
Entrez Gene ID
293117
Gene Name
ankyrin repeat domain 42
Gene Aliases
RGD1310789
Chromosome Location
Chr. 1: 157419117 - 157461588 on Build Rnor_6.0
UniGene ID
Rn.51262
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001135013.1 NP_001128485.1 1 502

View Full Product Details