Gene Transcripts

Gene Symbol
Olr83
Entrez Gene ID
293226
Gene Name
olfactory receptor 83
Gene Aliases
ratchr1-161034757-161035695_ORF
Chromosome Location
Chr. 1: 168326922 - 168327860 on Build Rnor_6.0
UniGene ID
Rn.141903
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001000136.1 NP_001000136.1 1 812

Gene Symbol
Olr81
Entrez Gene ID
293224
Gene Name
olfactory receptor 81
Gene Aliases
Chromosome Location
Chr. 1: 168293741 - 168294679 on Build Rnor_6.0
UniGene ID
Rn.141905
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001001271.1 NP_001001271.1 1 812

View Full Product Details