Gene Transcripts

Gene Symbol
Ric8a
Entrez Gene ID
293614
Gene Name
RIC8 guanine nucleotide exchange factor A
Gene Aliases
Ric-8A, Ric8
Chromosome Location
Chr. 1: 213606599 - 213612905 on Build Rnor_6.0
UniGene ID
Rn.94796
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001100520.1 NP_001093990.1 5 1118
GenBank mRNA AY177754.1 - 5 878
BC160852.1 - 5 1118 8364542
FQ223245.1 - 813

View Full Product Details