Gene Transcripts

Gene Symbol
Doc2g
Entrez Gene ID
293654
Gene Name
double C2-like domains, gamma
Gene Aliases
Doc2gp
Chromosome Location
Chr. 1: 219249247 - 219253790 on Build Rnor_6.0
UniGene ID
Rn.98629
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001011937.1 NP_001011937.1 8, 9 996
XM_006230735.2 XP_006230797.1 8, 9 2020
GenBank mRNA BC083779.1 - 8, 9 996

View Full Product Details