Gene Transcripts

Gene Symbol
Lage3
Entrez Gene ID
293863
Gene Name
L antigen family, member 3
Gene Aliases
RGD1562476
Chromosome Location
Chr. X: 156355376 - 156356799 on Build Rnor_6.0
UniGene ID
Rn.13531
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106345.1 NP_001099815.1 1 281

Gene Symbol
RGD1309036
Entrez Gene ID
292874
Gene Name
hypothetical LOC292874
Gene Aliases
Chromosome Location
Chr. 1: 100346096 - 100360635 on Build Rnor_6.0
UniGene ID
Rn.23075
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006228990.2 XP_006229052.1 2 598

View Full Product Details