Gene Transcripts

Gene Symbol
Pax2
Entrez Gene ID
293992
Gene Name
paired box 2
Gene Aliases
Chromosome Location
Chr. 1: 264493579 - 264583883 on Build Rnor_6.0
UniGene ID
Rn.137112
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106361.1 NP_001099831.1 5 625
XM_006231443.2 XP_006231505.1 6 1343
XM_008760403.1 XP_008758625.1 6 1340
XM_008760404.1 XP_008758626.1 6 1343
XM_008760405.1 XP_008758627.1 6 1340

View Full Product Details