Gene Transcripts

Gene Symbol
VSNL1
Entrez Gene ID
7447
Gene Name
visinin like 1
Gene Aliases
HLP3, HPCAL3, HUVISL1, VILIP, VILIP-1
Chromosome Location
Chr. 2: 17540540 - 17656439 on Build GRCh38
UniGene ID
Hs.444212
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_003385.4 NP_003376.2 3, 4 770
GenBank mRNA AB001104.1 - 3, 4 525
AF039555.1 - 3, 4 578
AK308816.1 - 636
BC022012.2 - 3, 4 607 4823954
U14747.1 - 3, 4 593

View Full Product Details