Gene Transcripts

Gene Symbol
Taar7g
Entrez Gene ID
294131
Gene Name
trace amine-associated receptor 7g
Gene Aliases
Ta9
Chromosome Location
Chr. 1: 22465092 - 22466168 on Build Rnor_6.0
UniGene ID
Rn.138143
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175588.1 NP_783178.1 1 427

View Full Product Details