Gene Transcripts

Gene Symbol
Mtch1
Entrez Gene ID
294313
Gene Name
mitochondrial carrier 1
Gene Aliases
Chromosome Location
Chr. 20: 6937341 - 6961165 on Build Rnor_6.0
UniGene ID
Rn.98516
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001100833.1 NP_001094303.1 7 738
XM_006256184.1 XP_006256246.1 7 760
GenBank mRNA BC098813.1 - 2 66
BC158661.1 - 7 738 8369713

View Full Product Details